Assay Details
| Gene Symbol: Milr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073165.1 | 5 - 6 | 701 | 90 | ||
| GenBank mRNA | AK137500.1 | - | 5 - 6 | 660 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
polymerase (DNA directed), gamma 2, accessory subunit |
Polg2 | Anti-sense | Chr. 11 - 106754821 - 106780010 on NCBI Build GRCm38 | ||
| Downstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 5 | Ddx5 |
2600009A06Rik, G17P1, HUMP68, Hlr1, p68 |
18.3kb | Anti-sense | Chr. 11 - 106780355 - 106789268 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Milr1
Genomic Map