Assay Details
| Gene Symbol: Socs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007707.3 | NP_031733.1 | 2 | 2205 | 101 | |
| GenBank mRNA | AK047165.1 | - | 2 | 2025 | 101 | |
| AK139241.1 | - | 2 | 2026 | 101 | ||
| AK150719.1 | - | 2 | 2025 | 101 | ||
| AK151761.1 | - | 2 | 2025 | 101 | ||
| AK152441.1 | - | 2 | 2025 | 101 | ||
| AK152468.1 | - | 2 | 2025 | 101 | ||
| AK152514.1 | - | 2 | 2025 | 101 | ||
| AK152686.1 | - | 2 | 2025 | 101 | ||
| AK157708.1 | - | 2 | 2011 | 101 | ||
| AK159395.1 | - | 2 | 2023 | 101 | ||
| AK170406.1 | - | 2 | 2023 | 101 | ||
| AK172399.1 | - | 2 | 2025 | 101 | ||
| BC052031.1 | - | 2 | 2001 | 5707539 | 101 | |
| U88328.1 | - | 1 | 1643 | 102 | ||
More Information
Set Membership: |
|

Socs3
Genomic Map