Assay Details
| Gene Symbol: RHOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127286.1 | NP_001120758.1 | 1 | 524 | 68 | |
| XM_012179330.3 | XP_012034720.1 | 3 | 1144 | 68 | ||
| XM_012179331.3 | XP_012034721.1 | 3 | 1132 | 68 | ||
| XM_012179334.3 | XP_012034724.1 | 3 | 1110 | 68 | ||
| GenBank mRNA | EU626206.1 | - | 1 | 524 | 68 | |
More Information
Set Membership: |
|

RHOH
Genomic Map