Assay Details
| Gene Symbol: POLR2K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011812.3 | XP_004011861.3 | 1 - 2 | 86 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004011812.3 transcripts.
Set Membership: |
|

POLR2K
Genomic Map