Assay Details
| Gene Symbol: SPINT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010455.4 | XP_004010504.2 | 7 - 8 | 1389 | 74 | |
| XM_015096948.2 | XP_014952434.1 | 7 - 8 | 1274 | 74 | ||
| XM_015096949.2 | XP_014952435.1 | 7 - 8 | 1352 | 74 | ||
More Information
Set Membership: |
|

SPINT1
Genomic Map