Assay Details
| Gene Symbol: CPLANE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965677.1 | XP_027821478.1 | 4 - 5 | 659 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPLANE2| Gene Symbol: CPLANE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965677.1 | XP_027821478.1 | 4 - 5 | 659 | 92 | |
Set Membership: |
|