Assay Details
| Gene Symbol: LOC101110127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963950.1 | XP_027819751.1 | 1 - 2 | 1070 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101110127| Gene Symbol: LOC101110127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963950.1 | XP_027819751.1 | 1 - 2 | 1070 | 71 | |
Set Membership: |
|