Assay Details
| Gene Symbol: LOC114110605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961584.1 | XP_027817385.1 | 3 - 4 | 633 | 93 | |
| Gene Symbol: LOC114110588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961529.1 | XP_027817330.1 | 3 - 4 | 447 | 93 | |
More Information
Set Membership: |
|

LOC114110605
Genomic Map