Assay Details
| Gene Symbol: RHOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275237.2 | XP_008273459.1 | 2 - 3 | 420 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008275237.2 transcripts.
Set Membership: |
|

RHOH
Genomic Map