Assay Details
| Gene Symbol: SPINT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269180.2 | XP_008267402.1 | 9 - 10 | 1762 | 63 | |
| XM_008269181.2 | XP_008267403.1 | 9 - 10 | 1675 | 63 | ||
| XM_008269182.2 | XP_008267404.1 | 10 - 11 | 1655 | 63 | ||
| XM_017348138.1 | XP_017203627.1 | 8 - 9 | 1637 | 63 | ||
More Information
Set Membership: |
|

SPINT1
Genomic Map