Assay Details
| Gene Symbol: CPLANE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722334.3 | XP_002722380.1 | 541 | 62 | ||
| XM_017338275.1 | XP_017193764.1 | 637 | 62 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPLANE2| Gene Symbol: CPLANE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722334.3 | XP_002722380.1 | 541 | 62 | ||
| XM_017338275.1 | XP_017193764.1 | 637 | 62 | |||
Set Membership: |
|