Assay Details
| Gene Symbol: TMPRSS11F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267796.2 | XP_008266018.1 | 9 - 10 | 1228 | 72 | |
| XM_017347521.1 | XP_017203010.1 | 8 - 9 | 1241 | 72 | ||
More Information
Set Membership: |
|

TMPRSS11F
Genomic Map