Assay Details
| Gene Symbol: LOC4330882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770701.2 | XP_015626187.1 | 1 - 2 | 179 | 89 | |
| GenBank mRNA | AK100497.1 | - | 1 - 2 | 68 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770701.2 transcripts.
Set Membership: |
|

LOC4330882
Genomic Map