Assay Details
| Gene Symbol: LOC4330882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770701.2 | XP_015626187.1 | 6 - 7 | 1116 | 121 | |
| GenBank mRNA | AK100497.1 | - | 6 - 7 | 1005 | 121 | |
More Information
Set Membership: |
|

LOC4330882
Genomic Map