Assay Details
| Gene Symbol: LOC4343220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791247.2 | XP_015646733.1 | 6 - 7 | 665 | 85 | |
| XM_015791248.2 | XP_015646734.1 | 5 - 6 | 927 | 85 | ||
| XM_015791249.2 | XP_015646735.1 | 6 - 7 | 621 | 85 | ||
| XM_015791250.2 | XP_015646736.1 | 6 - 7 | 660 | 85 | ||
| XM_015791251.2 | XP_015646737.1 | 6 - 7 | 616 | 85 | ||
| GenBank mRNA | AK059952.1 | - | 6 - 7 | 613 | 85 | |
More Information
Set Membership: |
|

LOC4343220
Genomic Map