Assay Details
| Gene Symbol: LOC4340511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788534.2 | XP_015644020.1 | 1 - 2 | 1276 | 88 | |
| GenBank mRNA | AK072030.1 | - | 1 - 2 | 1251 | 88 | |
| AK100405.1 | - | 1 - 2 | 1251 | 88 | ||
More Information
Set Membership: |
|

LOC4340511
Genomic Map