Assay Details
| Gene Symbol: LOC4349092 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759363.2 | XP_015614849.1 | 2 - 3 | 465 | 60 | |
| GenBank mRNA | AK070496.1 | - | 2 - 3 | 444 | 60 | |
More Information
Set Membership: |
|

LOC4349092
Genomic Map