Assay Details
| Gene Symbol: LOC4349092 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759363.2 | XP_015614849.1 | 6 - 7 | 975 | 65 | |
| GenBank mRNA | AK070496.1 | - | 6 - 7 | 954 | 65 | |
More Information
Set Membership: |
|

LOC4349092
Genomic Map