Assay Details
| Gene Symbol: LOC4327706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771264.2 | XP_015626750.1 | 2 - 3 | 407 | 67 | |
| XM_015771272.2 | XP_015626758.1 | 3 - 4 | 410 | 67 | ||
| GenBank mRNA | AB111915.1 | - | 1 - 2 | 258 | 67 | |
| AK058837.1 | - | 2 - 3 | 361 | 67 | ||
More Information
Set Membership: |
|

LOC4327706
Genomic Map