Assay Details
| Gene Symbol: LOC9271041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780609.2 | XP_015636095.1 | 1 - 2 | 606 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9271041| Gene Symbol: LOC9271041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780609.2 | XP_015636095.1 | 1 - 2 | 606 | 77 | |
Set Membership: |
|