Assay Details
| Gene Symbol: LOC4324936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757863.1 | XP_015613349.1 | 10 - 11 | 2416 | 109 | |
| GenBank mRNA | AK067564.1 | - | 10 - 11 | 2410 | 109 | |
| AK072312.1 | - | 10 - 11 | 2407 | 109 | ||
| AK103428.1 | - | 10 - 11 | 1861 | 109 | ||
More Information
Set Membership: |
|

LOC4324936
Genomic Map