Assay Details
| Gene Symbol: LOC4324936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757863.1 | XP_015613349.1 | 12 - 13 | 2890 | 76 | |
| GenBank mRNA | AK067564.1 | - | 12 - 13 | 2884 | 76 | |
More Information
Set Membership: |
|

LOC4324936
Genomic Map