Assay Details
| Gene Symbol: LOC4324936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757863.1 | XP_015613349.1 | 8 - 9 | 2121 | 90 | |
| GenBank mRNA | AK067564.1 | - | 8 - 9 | 2114 | 91 | |
| AK072312.1 | - | 8 - 9 | 2112 | 90 | ||
| AK103428.1 | - | 8 - 9 | 1566 | 90 | ||
More Information
Set Membership: |
|

LOC4324936
Genomic Map