Assay Details
Gene Symbol: LOC4332786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775992.2 | XP_015631478.1 | 1 - 2 | 187 | 133 | |
XM_015775993.2 | XP_015631479.1 | 1 - 2 | 188 | 133 | ||
GenBank mRNA | AK062464.1 | - | 1 - 2 | 180 | 133 | |
AK066021.1 | - | 1 - 2 | 184 | 133 | ||
AK066541.1 | - | 1 - 2 | 184 | 133 | ||
AK068987.1 | - | 1 - 2 | 175 | 133 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015775992.2, XM_015775993.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
