Assay Details
Gene Symbol: LOC4338944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783899.2 | XP_015639385.1 | 16 - 17 | 5422 | 93 | |
XM_015783900.2 | XP_015639386.1 | 16 - 17 | 5419 | 93 |
Gene Symbol: LOC4338940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784881.2 | XP_015640367.1 | 16 - 17 | 5453 | 93 | |
XM_015784882.2 | XP_015640368.1 | 16 - 17 | 5450 | 93 | ||
GenBank mRNA | AK103871.1 | - | 10 - 11 | 1074 | 93 | |
FJ536320.1 | - | 15 - 16 | 5090 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
