Assay Details
| Gene Symbol: LOC4338944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783899.2 | XP_015639385.1 | 17 - 18 | 5484 | 84 | |
| XM_015783900.2 | XP_015639386.1 | 17 - 18 | 5481 | 84 | ||
| Gene Symbol: LOC4338940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784881.2 | XP_015640367.1 | 17 - 18 | 5515 | 84 | |
| XM_015784882.2 | XP_015640368.1 | 17 - 18 | 5512 | 84 | ||
| XM_015784883.2 | XP_015640369.1 | 16 - 17 | 5381 | 84 | ||
| GenBank mRNA | AK103871.1 | - | 11 - 12 | 1136 | 84 | |
| FJ536320.1 | - | 16 - 17 | 5152 | 84 | ||
More Information
Set Membership: |
|

LOC4338944
Genomic Map