Assay Details
| Gene Symbol: LOC4338944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783899.2 | XP_015639385.1 | 8 - 9 | 4464 | 81 | |
| XM_015783900.2 | XP_015639386.1 | 8 - 9 | 4461 | 81 | ||
| XM_015783901.2 | XP_015639387.1 | 8 - 9 | 4468 | 81 | ||
| XM_015783902.2 | XP_015639388.1 | 8 - 9 | 4468 | 81 | ||
| Gene Symbol: LOC4338940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784881.2 | XP_015640367.1 | 8 - 9 | 4495 | 81 | |
| XM_015784882.2 | XP_015640368.1 | 8 - 9 | 4492 | 81 | ||
| XM_015784883.2 | XP_015640369.1 | 8 - 9 | 4501 | 81 | ||
| XM_015784884.2 | XP_015640370.1 | 8 - 9 | 4504 | 81 | ||
| GenBank mRNA | AK103871.1 | - | 2 - 3 | 116 | 81 | |
| FJ536320.1 | - | 7 - 8 | 4132 | 81 | ||
More Information
Set Membership: |
|

LOC4338944
Genomic Map