Assay Details
| Gene Symbol: LOC4326813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766610.2 | XP_015622096.1 | 4 - 5 | 819 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4326813| Gene Symbol: LOC4326813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766610.2 | XP_015622096.1 | 4 - 5 | 819 | 98 | |
Set Membership: |
|