Assay Details
| Gene Symbol: LOC9271041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780609.2 | XP_015636095.1 | 6 - 7 | 1573 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9271041| Gene Symbol: LOC9271041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780609.2 | XP_015636095.1 | 6 - 7 | 1573 | 80 | |
Set Membership: |
|