Assay Details
| Gene Symbol: LOC4342382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791972.2 | XP_015647458.1 | 1 - 2 | 425 | 84 | |
| GenBank mRNA | AK067910.1 | - | 1 - 2 | 408 | 84 | |
| AK287865.1 | - | 307 | 84 | |||
More Information
Set Membership: |
|

LOC4342382
Genomic Map