Assay Details
| Gene Symbol: LOC4342382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791972.2 | XP_015647458.1 | 7 - 8 | 984 | 135 | |
| GenBank mRNA | AK067910.1 | - | 7 - 8 | 967 | 135 | |
| AK287865.1 | - | 866 | 135 | |||
More Information
Set Membership: |
|

LOC4342382
Genomic Map