Assay Details
| Gene Symbol: CCM2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149270.2 | XP_015004756.2 | 274 | 63 | ||
| XM_028828294.1 | XP_028684127.1 | 274 | 63 | |||
| XM_028828295.1 | XP_028684128.1 | 386 | 63 | |||
| XM_028828296.1 | XP_028684129.1 | 409 | 63 | |||
| GenBank mRNA | JV571816.1 | - | 178 | 63 | ||
More Information
Set Membership: |
|

CCM2L
Genomic Map