Assay Details
| Gene Symbol: ISCU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261474.1 | NP_001248403.1 | 390 | 86 | ||
| GenBank mRNA | JU333707.1 | - | 390 | 86 | ||
| JV576687.1 | - | 430 | 86 | |||
More Information
Set Membership: |
|

ISCU
Genomic Map