Assay Details
| Gene Symbol: CPLANE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120871.2 | XP_014976357.1 | 848 | 138 | ||
| XM_015120913.2 | XP_014976399.1 | 582 | 138 | |||
| XM_028839153.1 | XP_028694986.1 | 315 | 138 | |||
More Information
Set Membership: |
|

CPLANE2
Genomic Map