Assay Details
| Gene Symbol: PLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266703.1 | NP_001253632.1 | 232 | 133 | ||
| XM_028853380.1 | XP_028709213.1 | 542 | 133 | |||
| XM_028853382.1 | XP_028709215.1 | 109 | 133 | |||
| GenBank mRNA | DQ148090.1 | - | 194 | 133 | ||
| JU333277.1 | - | 236 | 133 | |||
| JV045160.1 | - | 232 | 133 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028853382.1 transcripts.
Set Membership: |
|

PLPP4
Genomic Map