Assay Details
| Gene Symbol: POLR2K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145794.2 | XP_015001280.1 | 589 | 86 | ||
| XM_015145795.2 | XP_015001281.1 | 204 | 86 | |||
| XM_015145796.2 | XP_015001282.1 | 219 | 86 | |||
| XM_028852798.1 | XP_028708631.1 | 206 | 86 | |||
| GenBank mRNA | JU473271.1 | - | 136 | 86 | ||
| JV567594.1 | - | 149 | 86 | |||
| JV639047.1 | - | 178 | 86 | |||
More Information
Set Membership: |
|

POLR2K
Genomic Map