Assay Details
| Gene Symbol: PRSS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266588.1 | NP_001253517.1 | 557 | 108 | ||
| XM_015125992.2 | XP_014981478.1 | 997 | 108 | |||
| GenBank mRNA | JU476439.1 | - | 557 | 108 | ||
More Information
Set Membership: |
|

PRSS8
Genomic Map