Assay Details
| Gene Symbol: CLPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260887.1 | NP_001247816.1 | 307 | 87 | ||
| GenBank mRNA | JU332619.1 | - | 308 | 87 | ||
| JU474082.1 | - | 321 | 87 | |||
| JV045243.1 | - | 307 | 87 | |||
More Information
Set Membership: |
|

CLPX
Genomic Map