Assay Details
| Gene Symbol: MRPL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260951.1 | NP_001247880.1 | 567 | 73 | ||
| GenBank mRNA | JU332899.1 | - | 554 | 73 | ||
| JU474289.1 | - | 554 | 73 | |||
| JV046171.1 | - | 567 | 73 | |||
More Information
Set Membership: |
|

MRPL10
Genomic Map