Assay Details
| Gene Symbol: RPL7L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193633.1 | NP_001180562.1 | 948 | 117 | ||
| GenBank mRNA | CN648780.1 | - | 948 | 117 | ||
| JU334109.1 | - | 931 | 117 | |||
| JV046816.1 | - | 938 | 117 | |||
| JV046817.1 | - | 938 | 117 | |||
| JV046818.1 | - | 938 | 117 | |||
| JV046819.1 | - | 938 | 117 | |||
More Information
Set Membership: |
|

RPL7L1
Genomic Map