Assay Details
| Gene Symbol: MILR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120115.2 | XP_014975601.1 | 784 | 82 | ||
| XM_028836669.1 | XP_028692502.1 | 524 | 82 | |||
| GenBank mRNA | JU471204.1 | - | 716 | 82 | ||
| JV704559.1 | - | 715 | 82 | |||
More Information
Set Membership: |
|

MILR1
Genomic Map