Assay Details
| Gene Symbol: Crlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134465.2 | NP_604460.2 | 3 - 4 | 466 | 73 | |
| GenBank mRNA | AF404510.1 | - | 3 - 4 | 409 | 73 | |
| BC078740.1 | - | 3 - 4 | 466 | 7108137 | 73 | |
| FQ232217.1 | - | 3 - 4 | 452 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crlf2
Genomic Map