Assay Details
| Gene Symbol: Prss8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138836.1 | NP_620191.1 | 2 - 3 | 329 | 65 | |
| GenBank mRNA | AB017638.1 | - | 2 - 3 | 329 | 65 | |
| AF202076.1 | - | 2 - 3 | 327 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prss8
Genomic Map