Assay Details
| Gene Symbol: Ap2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022952.2 | NP_075241.2 | 1 - 2 | 69 | 87 | |
| GenBank mRNA | BC088138.1 | - | 1 - 2 | 69 | 7374432 | 87 |
| FQ234449.1 | - | 1 - 2 | 88 | 87 | ||
| U75917.1 | - | 1 - 2 | 88 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC088138.1,NM_022952.2,U75917.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ap2s1
Genomic Map