Assay Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047871.1 | NP_001041336.1 | 1 - 2 | 289 | 81 | |
| GenBank mRNA | BC161810.1 | - | 1 - 2 | 223 | 9023096 | 81 |
| DQ480751.1 | - | 1 - 2 | 289 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifrd2
Genomic Map