Assay Details
| Gene Symbol: Polr3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107784.1 | NP_001101254.1 | 7 - 8 | 886 | 111 | |
| XM_008762269.1 | XP_008760491.1 | 7 - 8 | 917 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polr3f
Genomic Map