Assay Details
| Gene Symbol: Clpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007803.1 | NP_001007804.1 | 13 - 14 | 1809 | 106 | |
| XM_006243250.1 | XP_006243312.1 | 12 - 13 | 2012 | 106 | ||
| GenBank mRNA | BC085867.1 | - | 13 - 14 | 1935 | 7190926 | 106 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
biological_process
positive regulation of peptidase activity
proteolysis involved in cellular protein catabolic process
molecular_function
ATP-dependent peptidase activity
ATP binding
peptidase activator activity
ATPase activity
metal ion binding
unfolded protein binding
protein folding
biological_process
positive regulation of peptidase activity
proteolysis involved in cellular protein catabolic process
molecular_function
ATP-dependent peptidase activity
ATP binding
peptidase activator activity
ATPase activity
metal ion binding
unfolded protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P026339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154699 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026339 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026339 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P026339 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16842 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53128 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35013 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4910 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10910952 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910951 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910943 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910950 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910939 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910941 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910948 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910942 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910947 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910937 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910940 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910949 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910938 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910944 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910946 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910945 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10910936 | - | - |
| Rat Genome 230 2.0 Array | 1398698_at | - | - |
| Rat Genome 230 2.0 Array | 1377192_a_at | - | - |
| Rat Genome 230 2.0 Array | 1398698_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377192_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1377192_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1377192_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800006760.rn.1 | TC0800000937.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800006761.rn.1 | TC0800000937.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800006763.rn.1 | TC0800000937.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800006762.rn.1 | TC0800000937.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800006758.rn.1 | TC0800000937.rn.1 | RTA TC |
| Rat Expression Set 230 | 1377192_a_at | - | - |
| Rat Expression Set 230 | 1377192_PM_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910936 | - | - |
| Rat Genome U34 Set | rc_AI029291_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21540279 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040542 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1530376 | - | - |
| RatRef-12 Expression BeadChip | ILMN_63961 | - | - |
| RatRef-12 Expression BeadChip | ILMN_160625 | - | - |

Clpx
Genomic Map