Assay Details
| Gene Symbol: Tmprss13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127528.1 | NP_001121000.1 | 7 - 8 | 884 | 63 | |
| GenBank mRNA | BC166801.1 | - | 7 - 8 | 884 | 7938493 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss13
Genomic Map