Assay Details
| Gene Symbol: Tek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105737.1 | NP_001099207.1 | 22 - 23 | 3610 | 123 | |
| XM_008763910.1 | XP_008762132.1 | 22 - 23 | 3430 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
vasculogenesis
response to hypoxia
positive regulation of protein phosphorylation
endothelial cell proliferation
endochondral ossification
sprouting angiogenesis
positive regulation of cytokine secretion involved in immune response
cell-matrix adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
heart development
response to organic substance
positive regulation of endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase signaling
single organismal cell-cell adhesion
negative regulation of angiogenesis
peptidyl-tyrosine phosphorylation
signal transduction by protein phosphorylation
hemopoiesis
regulation of cell migration
organ regeneration
response to retinoic acid
regulation of establishment or maintenance of cell polarity
positive regulation of peptidyl-serine phosphorylation
substrate adhesion-dependent cell spreading
intracellular signal transduction
positive regulation of protein import into nucleus
negative regulation of apoptotic process
response to peptide hormone
positive regulation of phosphatidylinositol 3-kinase activity
response to estrogen
regulation of angiogenesis
positive regulation of angiogenesis
positive regulation of cell adhesion
protein autophosphorylation
Tie signaling pathway
protein oligomerization
response to cAMP
positive regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
heart trabecula formation
positive regulation of ERK1 and ERK2 cascade
regulation of NIK/NF-kappaB signaling
positive regulation of intracellular signal transduction
positive regulation of actin cytoskeleton reorganization
negative regulation of endothelial cell apoptotic process
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
receptor activity
ATP binding
growth factor binding
angiogenesis
vasculogenesis
response to hypoxia
positive regulation of protein phosphorylation
endothelial cell proliferation
endochondral ossification
sprouting angiogenesis
positive regulation of cytokine secretion involved in immune response
cell-matrix adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
heart development
response to organic substance
positive regulation of endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase signaling
single organismal cell-cell adhesion
negative regulation of angiogenesis
peptidyl-tyrosine phosphorylation
signal transduction by protein phosphorylation
hemopoiesis
regulation of cell migration
organ regeneration
response to retinoic acid
regulation of establishment or maintenance of cell polarity
positive regulation of peptidyl-serine phosphorylation
substrate adhesion-dependent cell spreading
intracellular signal transduction
positive regulation of protein import into nucleus
negative regulation of apoptotic process
response to peptide hormone
positive regulation of phosphatidylinositol 3-kinase activity
response to estrogen
regulation of angiogenesis
positive regulation of angiogenesis
positive regulation of cell adhesion
protein autophosphorylation
Tie signaling pathway
protein oligomerization
response to cAMP
positive regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
heart trabecula formation
positive regulation of ERK1 and ERK2 cascade
regulation of NIK/NF-kappaB signaling
positive regulation of intracellular signal transduction
positive regulation of actin cytoskeleton reorganization
negative regulation of endothelial cell apoptotic process
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
receptor activity
ATP binding
growth factor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P534601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107354 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P534601 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P534601 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P534601 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P534601 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P335594 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 105025 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81080 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84248 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92365 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101912 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10869966 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869959 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869954 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869948 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869950 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869962 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869953 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869967 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869965 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869949 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869958 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869960 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869955 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869961 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869947 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869951 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869956 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869957 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869963 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869964 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869952 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10869946 | - | - |
| Rat Genome 230 2.0 Array | 1393067_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393067_at | - | - |
| Affymetrix HT Rat Focus Array | 1393067_at | - | - |
| Affymetrix HT Rat Focus Array | 1393067_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500006266.rn.1 | TC0500000993.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393067_PM_at | - | - |
| Rat Expression Set 230 | 1393067_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10869946 | - | - |
| Rat Genome U34 Set | rc_AI058402_at | - | - |
| Rat Genome U34 Set | rc_AA957010_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22330185 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370024 | - | - |

Tek
Genomic Map