Assay Details
| Gene Symbol: LOC100910205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750592.3 | XP_003750640.3 | 4 - 5 | 1090 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100910205| Gene Symbol: LOC100910205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750592.3 | XP_003750640.3 | 4 - 5 | 1090 | 85 | |
Set Membership: |
|